Learn More
SCAT™ Fittings for HPLC Systems
Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.
151.00 SEK - 1065.00 SEK
Product Code | Brand | Color | For Use With | Includes | Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | For Use With | Includes | Type | Price | Quantity & Availability | ||
13570810
|
SCAT™
160502 |
Colorless | Capillary Connector for HPLC/UHPLC Systems | UNF 0.25 in. 28G Plug | Plug |
303.00 SEK
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10839150
|
SCAT™
107811 |
White | 2 to 3 mm ID Tubing | Straight Tube Connector (2 to 3 mm ID) | Connector |
151.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10282990
|
SCAT™
107812 |
White | 3 to 4 mm ID Tubing | Straight Tube Connector (3 to 4 mm ID) | Connector |
151.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10891341
|
SCAT™
107813 |
White | 4 to 6 mm ID Tubing | Straight Tube Connector (4 to 6 mm ID) | Connector |
151.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10890861
|
SCAT™
160143 |
White | 6.4 to 8 mm ID Tubing | Curved Tube Connector (6.4 to 8 mm ID) | Connector |
379.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10151263
|
SCAT™
107041 |
White | Safety Caps, Safety Waste Caps | Fitting with Integrated Ferrule (1.6 mm OD) | Fitting |
1065.00 SEK
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10691894
|
SCAT™
107043 |
White | Safety Caps, Safety Waste Caps | Fitting with Integrated Ferrule (3.2 mm OD) | Fitting |
1065.00 SEK
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
10593405
|
SCAT™
107045 |
White | Safety Caps, Safety Waste Caps | Fitting with Integrated Ferrule (4.76 mm OD, UNF 5/16 to 24 in.) | Fitting |
379.00 SEK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10773882
|
SCAT™
160142 |
White | Safety Waste Caps, HPLC and UHPLC Systems | Straight Tube Connector (6.4 to 8 mm ID) | Connector |
303.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||
10120883
|
SCAT™
107044 |
White | 6.35 mm Preparative HPLC Tube | Fitting with Integrated Ferrule (6.35 mm, NPT 1/8 in.) | Fitting |
354.00 SEK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||
10282320
|
SCAT™
107047 |
White | 4 mm Diameter Tube | Fitting with Integrated Ferrule (4 mm OD, UNF 5/16 to 24 in.) | Fitting |
379.00 SEK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||
17114274
|
SCAT™
117819 |
White | Corrugated Tube (8 mm ID, 11 mm OD) | Compression Fitting for Tube Connector, Tube Olive for Corrugated tube (8 mm ID, 11 mm OD), with Locknut | Fitting |
760.00 SEK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10671771
|
SCAT™
107063 |
Blue | Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems | Fitting with Integrated Ferrule (3.2 mm OD) | Fitting |
303.00 SEK
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
10795772
|
SCAT™
117808 |
Blue | Tube Connector | Curved Tube Connector (5 to 11.5 mm ID) | Connector |
181.00 SEK
Each |
In Stock
Log in to see stock. |
||
10202090
|
SCAT™
107061 |
Green | Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems | Fitting with Integrated Ferrule (1.6 mm OD) | Fitting |
303.00 SEK
Pack of 5 |
In Stock
Log in to see stock. |
||
10292320
|
SCAT™
107059 |
Violet | Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems | Fitting with Integrated Ferrule (2.3 mm OD) | Fitting |
303.00 SEK
Pack of 5 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||
14737170
|
SCAT™
107905 |
White | Capillary | Ferrule, 10 Pk. | Ferrule |
442.49 SEK
Pack of 10 |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
14747170
|
SCAT™
107906 |
White | Capillary | Ferrule, 10 Pk. | Ferrule |
442.49 SEK
Pack of 10 |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
14757170
|
SCAT™
107908 |
White | Capillary | Ferrule, 10 Pk. | Ferrule |
442.49 SEK
Pack of 10 |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17916970
|
SCAT™
160530 |
Red | Suitable for all SCAT PFA fittings. | Installation Wrench POM, Hexagonal | Wrench |
294.50 SEK
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10509490
|
SCAT™
160500 |
Silver | PFA Fittings | Hexagonal Installation Wrench, CleanCut Capillary Cutter, Replacement Blade | Installation Wrench |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10715572
|
SCAT™
107066 |
Colorless | Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems | Fitting with Integrated Ferrule (3.2 mm OD) | Fitting |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15491793
|
SCAT™
107048 |
Colorless | Capillary (1.6 mm OD) | Fitting with Integrated Ferrule (1.6 mm OD) | Fitting |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10510631
|
SCAT™
160499 |
Black | Polypropylene Fittings (e.g. Earlier Generation of Safety Caps) | Square Installation Wrench | Installation Wrench |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10303970
|
SCAT™
107060 |
Black | Capillary (3.2 mm OD) | Fitting with Integrated Ferrule (3.2 mm OD) | Fitting |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
SCAT fittings are available in the following versions:
- PFA fitting with integrated ferrule in various colors and diameters
- Tube connectors in various diameters