Learn More
SCAT™ PTFE Thread Adapters
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand: SCAT™
798.73 SEK valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polytetrafluoroethylene (PTFE) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifications
Thread Adapter | |
PTFE | |
GL32 (f) to GL45 (m) |