Learn More
IKA™ T 50 Digital ULTRA-TURRAX™ Disperser
GREENER_CHOICE
Offering a wide speed range from 500 to 10,000rpm that enables users to work at high circumferential speeds even with small rotor diameters.
Brand:  IKA™ 0003787000
Product Code. 13306309
  • 47765.00 SEK / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Powerful dispersing instrument for volumes from 0.25 to 30L (H2O) with digital speed display
  • Broad choice of dispersing elements guarantees a wide range of applications
  • Reproducible operations due to constant speed even with changes in viscosities
  • Electronic speed control
  • Electronic overload protection
  • Stainless steel dispersing elements can be cleaned quickly and easily
  • Smooth start
  • Error code display
Specifications
Specifications
Digital Disperser
220/240 V
50/60 Hz
250 mL to 30 L
355 mm
LED
T 50 ULTRA-TURRAX™
5000 mPas
600 to 10,000 rpm
220/240 V, 50/60 Hz, 1100 W
1100 W
139 mm
115 mm
Dispersing at high circumferential speeds with small rotor diameters
1 Ea.
5.76 kg