Learn More
SCAT Waste Disposal Set, V2.0
The SCAT Waste Disposal Sets 1-4 are combinations of funnels and canisters for the safe disposal of liquid laboratory waste.
Brand: SCAT 307320
2695.25 SEK valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 4.35800kg
Description
- Made of high quality and electrically conductive HDPE material
- With grounding cable to dissipate the electrical charge
- With splash guard and removable sieve for safe filling
- Sets consist of ball or lid funnels for individual applications
- 10 or 20 L disposal canisters with mechanical level indicators
- Can be optionally combined with collection tray
All canisters and funnels are connected via an S60/61 thread and their components are perfectly matched to each other.
Specifications
Funnel with ball Arnold, V2.0, S60/61, PE-HD electrostatic conductive, with lance (220 mm), splash guard, removable sieve and earthing cable (317621), 10 L canister, S60/61, with floater, HDPE electrostatic conductive (108042) | |
Set 1 | |
HPDE Electrostatic Conductive | |
S60/61 |