man

Test

CAS RN 100-62-9

H2CN

CAS RN 100-62-9

IUPAC Name: N-phenylmethanimine
Synonyms: N-phenylmethanimine
Molecular Weight (g/mol): 105.14
Molecular Formula: C7H7N
InChi Key: VULHYRAYXYTONQ-UHFFFAOYSA-N
SMILES: C=NC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results