man

Test

CAS RN 105228-46-4

OONOO

CAS RN 105228-46-4

IUPAC Name: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Synonyms: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol): 387.44
Molecular Formula: C24H21NO4
InChi Key: HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)