CAS RN 107534-96-3
CAS RN 107534-96-3
IUPAC Name:
1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol
Synonyms:
tebuconazole
Molecular Weight (g/mol):
307.82
Molecular Formula:
C16H22ClN3O
InChi Key:
PXMNMQRDXWABCY-UHFFFAOYNA-N
SMILES:
CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1
1
–
7
of
7
results
4
Thermo Scientific Chemicals Tebuconazole
CAS: 107534-96-3 Molecular Formula: C16H22ClN3O Molecular Weight (g/mol): 307.82 MDL Number: MFCD02674797 InChI Key: PXMNMQRDXWABCY-UHFFFAOYNA-N IUPAC Name: 1-(4-chlorophenyl)-4,4-dimethyl-3-[(1H-1,2,4-triazol-1-yl)methyl]pentan-3-ol SMILES: CC(C)(C)C(O)(CCC1=CC=C(Cl)C=C1)CN1C=NC=N1
7
Tebuconazole, Fluka™
Analytical standard, reference material