CAS RN 109-00-2
CAS RN 109-00-2
IUPAC Name:
pyridin-3-ol
Synonyms:
2-pyridone
Molecular Weight (g/mol):
95.1
Molecular Formula:
C5H5NO
InChi Key:
GRFNBEZIAWKNCO-UHFFFAOYSA-N
SMILES:
OC1=CC=CN=C1
1
–
2
of
2
results
1
3-Hydroxypyridine, 98%, Thermo Scientific Chemicals
CAS: 109-00-2 Molecular Formula: C5H5NO Molecular Weight (g/mol): 95.101 MDL Number: MFCD00006378 InChI Key: GRFNBEZIAWKNCO-UHFFFAOYSA-N Synonym: 3-hydroxypyridine,3-pyridinol,3-pyridol,3-pyridone,3-oxopyridine,beta-hydroxypyridine,m-hydroxypyridine,3-pyridyl alcohol,3-hydoxypyridine,unii-4kbe4p5b6s PubChem CID: 7971 IUPAC Name: pyridin-3-ol SMILES: C1=CC(=CN=C1)O
2
3-Hydroxypyridine, 98%, Thermo Scientific Chemicals
CAS: 109-00-2 Molecular Formula: C5H5NO Molecular Weight (g/mol): 95.1 MDL Number: MFCD00006378 InChI Key: GRFNBEZIAWKNCO-UHFFFAOYSA-N Synonym: 3-hydroxypyridine,3-pyridinol,3-pyridol,3-pyridone,3-oxopyridine,beta-hydroxypyridine,m-hydroxypyridine,3-pyridyl alcohol,3-hydoxypyridine,unii-4kbe4p5b6s PubChem CID: 7971 IUPAC Name: pyridin-3-ol SMILES: C1=CC(=CN=C1)O