man

Test

CAS RN 121219-03-2

OHFBr

CAS RN 121219-03-2

IUPAC Name: 4-bromo-3-fluorophenol
Synonyms: 4-bromo-3-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: MRQYTJXVULSNIS-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(Br)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results