man

Test

CAS RN 123-09-1

H3CSCl

CAS RN 123-09-1

IUPAC Name: 1-chloro-4-(methylsulfanyl)benzene
Synonyms: 4-chlorophenyl methyl sulfide
Molecular Weight (g/mol): 158.64
Molecular Formula: C7H7ClS
InChi Key: KIQQUVJOLVCZKG-UHFFFAOYSA-N
SMILES: CSC1=CC=C(Cl)C=C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)