CAS RN 124-48-1
CAS RN 124-48-1
IUPAC Name:
dibromo(chloro)methane
Synonyms:
dibromochloromethane C-H-br2-cl chbr2cl ccris 938 CDBM
Molecular Weight (g/mol):
208.28
Molecular Formula:
CHBr2Cl
InChi Key:
GATVIKZLVQHOMN-UHFFFAOYSA-N
SMILES:
ClC(Br)Br
1
–
2
of
2
results
1
Dibromochloromethane, 97+%, Thermo Scientific Chemicals
CAS: 124-48-1 Molecular Formula: CHBr2Cl Molecular Weight (g/mol): 208.277 MDL Number: MFCD00000820 InChI Key: GATVIKZLVQHOMN-UHFFFAOYSA-N Synonym: chlorodibromomethane,methane, dibromochloro,dibromo chloro methane,monochlorodibromomethane,cdbm,methane, chlorodibromo,unii-3t4ajr1h24,ccris 938,dibromo-chloro-methane,chlorobromoform PubChem CID: 31296 ChEBI: CHEBI:34627 IUPAC Name: dibromo(chloro)methane SMILES: C(Cl)(Br)Br