CAS RN 139-33-3
CAS RN 139-33-3
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Solution ready to use, for volumetric analysis, Fisher Chemical™
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 IUPAC Name: disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA disodium salt solution, Volumetric, 0.05 M EDTA-Na2, Honeywell Fluka™
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical™
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic acid disodium salt, 0.100N (0.050M) Standardized solution, Thermo Scientific Chemicals
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 IUPAC Name: disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution, Thermo Scientific Chemicals
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 IUPAC Name: disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Promotions are available
Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution, Thermo Scientific Chemicals
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 IUPAC Name: disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry
Promotions are available
EDTA disodium salt concentrate, For 1L standard solution, 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L PubChem CID: 57339238 ChEBI: CHEBI:64734 IUPAC Name: disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Honeywell Fluka™
Chemical Name or Material: Ethylenediaminetetraacetic acid disodium salt solution CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 UN Number: NONH for all modes of transport Beilstein: 3822669 Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O Molecular Weight (g/mol): 336.21 PubChem CID: 57339238 ChEBI: CHEBI:64734
Ethylenediaminetetraacetic Acid Disodium Salt Solution volumetric, Honeywell Fluka™
CAS: 139-33-3 Molecular Formula: C10H14N2Na2O8 Molecular Weight (g/mol): 336.21 MDL Number: MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 InChI Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L Synonym: ethylenediaminetetraacetic acid disodium salt,edta na2,disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid PubChem CID: 57339238 ChEBI: CHEBI:64734 SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
- 1
- 2