CAS RN 52613-28-2
CAS RN 52613-28-2
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
Methyl deacetylasperulosidate, MedChemExpress
MedChemExpress Methyl deacetylasperulosidate is an iridoid and shows purgative effects in mice and lowers the blood glucose level in normal mice.