CAS RN 55297-96-6
CAS RN 55297-96-6
IUPAC Namn:
(2E)-but-2-enedioic acid; (2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate
Synonymer:
(2R,3S,4S,6R,8S,14R)-4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[2-(diethylamino)ethyl]sulfanyl}acetate; fumaric acid
Molekylvikt (g/mol):
609.82
Molekylär sammansättning:
C32H51NO8S
InChi Key:
YXQXDXAHCSEVSD-LKNSULBJSA-N
SMILES:
OC(=O)\C=C\C(O)=O.CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)[C@@H](O)[C@H](C)C23CCC(=O)[C@@H]2C1(C)[C@H](C)CC3
1
–
4
av
4
Resultat
2
Tiamulin Hydrogen Fumarate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Tiamulin Hydrogen Fumarate, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.