CAS RN 637-81-0
CAS RN 637-81-0
IUPAC Namn:
ethyl 2-azidoacetate
Synonymer:
ethyl 2-azidoacetate
Molekylvikt (g/mol):
129.12
Molekylär sammansättning:
C4H7N3O2
InChi Key:
HVJJYOAPXBPQQV-UHFFFAOYSA-N
SMILES:
CCOC(=O)CN=[N+]=[N-]
1
–
5
av
5
Resultat
3
Ethyl 2-Azidoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ethyl 2-Azidoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
Ethyl 2-Azidoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.