CAS RN 968-81-0
CAS RN 968-81-0
IUPAC Namn:
3-(4-acetylbenzenesulfonyl)-1-cyclohexylurea
Synonymer:
acetohexamide hypoglicil gamadiabet acetohexamid dymelor tsiklamid ordimel minoral metaglucina cyclamide
Molekylvikt (g/mol):
324.4
Molekylär sammansättning:
C15H20N2O4S
InChi Key:
VGZSUPCWNCWDAN-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
1
–
2
av
2
Resultat
1
Acetohexamide, LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.