missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-1,1-dichloroethane, 97%, Thermo Scientific™

Product Code. 11433048
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-81-0
Molecular Formula C2H2Br2Cl2
Molecular Weight (g/mol) 256.746
MDL Number MFCD00053228
InChI Key FIYBYNHDEOSJPL-UHFFFAOYSA-N
Synonym ethane, 1,2-dibromo-1,1-dichloro, 1,2-dibromo-2,2-dichloroethane, unii-8d6fn8f1wd, 8d6fn8f1wd, ethane, dibromodichloro, ethane,2-dibromo-1,1-dichloro, 1,2-dibromo-1,1-dichloroethane, 100 microg/ml in methanol
PubChem CID 6401
IUPAC Name 1,2-dibromo-1,1-dichloroethane
SMILES C(C(Cl)(Cl)Br)Br

Specifications

Quantity 50 g
UN Number UN2810
Formula Weight 256.75
Percent Purity 97%
Chemical Name or Material 1,2-Dibromo-1,1-dichloroethane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'