missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenoxy-2-propanol, tech. 85%, Thermo Scientific Chemicals
398.80 SEK - 1049.65 SEK
Chemical Identifiers
CAS | 770-35-4 |
---|---|
Molecular Formula | C9H12O2 |
Molecular Weight (g/mol) | 152.19 |
MDL Number | MFCD00016861 |
InChI Key | IBLKWZIFZMJLFL-UHFFFAOYNA-N |
Synonym | 1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312 |
PubChem CID | 92839 |
IUPAC Name | 1-phenoxypropan-2-ol |
SMILES | CC(O)COC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11331637
|
Thermo Scientific Alfa Aesar
L04596.18 |
50 g |
398.80 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341637
|
Thermo Scientific Alfa Aesar
L04596.30 |
250 g |
1049.65 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
770-35-4 | |
152.19 | |
IBLKWZIFZMJLFL-UHFFFAOYNA-N | |
92839 | |
CC(O)COC1=CC=CC=C1 |
C9H12O2 | |
MFCD00016861 | |
1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312 | |
1-phenoxypropan-2-ol |
Specifications
770-35-4 | |
242°C to 244°C | |
C9H12O2 | |
MFCD00016861 | |
1-phenoxy-2-propanol, 2-propanol, 1-phenoxy, phenoxyisopropanol, propylene phenoxetol, 2-phenoxy-1-methylethanol, 1-phenoxy-propan-2-ol, phenyl-.beta.-hydroxypropyl ether, dsstox_cid_7312, dsstox_rid_78402, dsstox_gsid_27312 | |
CC(O)COC1=CC=CC=C1 | |
152.19 | |
152.19 | |
Technical |
1.06 | |
120°C (248°F) | |
1.523 | |
1941356 | |
IBLKWZIFZMJLFL-UHFFFAOYNA-N | |
1-phenoxypropan-2-ol | |
92839 | |
85% | |
1-Phenoxy-2-propanol |
Safety and Handling
missing translation for 'einecsNumber' : 212-222-7
missing translation for 'rtecsNumber' : UB8886500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only