missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Amino-2-propyl)aniline, 95%, Thermo Scientific™

Product Code. 15427597
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 229326-17-4
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.23
MDL Number MFCD08276909
InChI Key MWUKUEYFFYQSQE-UHFFFAOYSA-N
Synonym 2-2-aminopropan-2-yl aniline, 2-2-amino-2-propyl aniline, ambkkkkk207, 2-2-amino-2-propanyl aniline, 2-2-aminophenyl-2-propylamine, 2-amino-alpha,alpha-dimethylbenzylamine, benzenemethanamine, 2-amino-alpha,alpha-dimethyl-9ci
PubChem CID 21719463
IUPAC Name 2-(2-aminopropan-2-yl)aniline
SMILES CC(C)(N)C1=CC=CC=C1N

Specifications

Quantity 1 g
UN Number UN2810
Formula Weight 150.22
Percent Purity 95%
Chemical Name or Material 2-(2-Amino-2-propyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'