missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorothiophenol, 97%, Thermo Scientific™

Product Code. 11472867
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17231-95-7
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041420
InChI Key QGRKONUHHGBHRB-UHFFFAOYSA-N
Synonym 2,3-dichlorothiophenol, 2,3-dichlorobenzene-1-thiol, dichlorothiophenol, benzenethiol, dichloro, 2,3-dichloro thiophenol, pubchem6796, 2,3-dichiorothiophenol, 2,3-dichloro-thiophenol, acmc-1cf9p, 2,3-dichlorobenzene thiol
PubChem CID 140202
IUPAC Name 2,3-dichlorobenzenethiol
SMILES C1=CC(=C(C(=C1)Cl)Cl)S

Specifications

Melting Point 55°C to 58°C
Boiling Point 95°C to 98°C (0.5 mmHg)
Odor Unpleasant
Quantity 5 g
Beilstein 2043616
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorothiophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'