Learn More
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%, Thermo Scientific Chemicals
430.75 SEK - 1254.38 SEK
Chemical Identifiers
CAS | 4628-94-8 |
---|---|
Molecular Formula | C2K2N2S3 |
Molecular Weight (g/mol) | 226.413 |
MDL Number | MFCD00003104 |
InChI Key | GPWLFGDMYSVEGN-UHFFFAOYSA-L |
Synonym | 2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole |
PubChem CID | 78364 |
IUPAC Name | dipotassium;1,3,4-thiadiazole-2,5-dithiolate |
SMILES | C1(=NN=C(S1)[S-])[S-].[K+].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11357556
|
Thermo Scientific Alfa Aesar
L11105.06 |
5 g |
430.75 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367556
|
Thermo Scientific Alfa Aesar
L11105.14 |
25 g |
1254.38 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
4628-94-8 | |
226.413 | |
GPWLFGDMYSVEGN-UHFFFAOYSA-L | |
78364 | |
C1(=NN=C(S1)[S-])[S-].[K+].[K+] |
C2K2N2S3 | |
MFCD00003104 | |
2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole | |
dipotassium;1,3,4-thiadiazole-2,5-dithiolate |
Specifications
4628-94-8 | |
C2K2N2S3 | |
4917786 | |
Insoluble in water. | |
C1(=NN=C(S1)[S-])[S-].[K+].[K+] | |
226.413 | |
226.43 | |
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt |
∽274°C (decomposition) | |
MFCD00003104 | |
2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole | |
GPWLFGDMYSVEGN-UHFFFAOYSA-L | |
dipotassium;1,3,4-thiadiazole-2,5-dithiolate | |
78364 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 225-043-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only