missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)oxetane, 96%, Thermo Scientific Chemicals

Product Code. 15472719
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939759-23-6
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.00
MDL Number MFCD08544405
InChI Key OMXAGUVERXNCSZ-UHFFFAOYNA-N
Synonym 2-bromomethyl oxetane, 2-bromomethyl-oxetane, oxetane, 2-bromomethyl, cyclobutane, bromomethyl, 2-bromomethyl oxetane oxetane, 2-bromomethyl
PubChem CID 16244494
IUPAC Name 2-(bromomethyl)oxetane
SMILES BrCC1CCO1

Specifications

Color Colorless to Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 151
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(Bromomethyl)oxetane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'