missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Anti- Trastuzumab (Anti-Idiotype) Antibody, R&D Systems™

Rabbit Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB95471-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Trastuzumab Monoclonal specifically detects Trastuzumab in Multi-Species samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifikationer
| Trastuzumab | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Full-length Trastuzumab | |
| 25 μg | |
| Detects Trastuzumab in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with Cetuximab, Ibritumomab, Rituximab, Adalimumab, Tositumomab, Brentuximab, Gemtuzumab, Solanezumab, Teprotumumab, and Aducanumab is observed. | |
| Multi-species | |
| Supernatant |
| Western Blot, Flow Cytometry, ELISA | |
| 2261E | |
| Western Blot 0.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, ELISA | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering