missing translation for 'onlineSavingsMsg'
Läs mer

3-Amino-2,6-dimethylpyridine, 97%, Thermo Scientific™

Produktkod. 11319835
Click to view available options
Kvantitet:
1 g
5 g
25 g
Tamaño de la unidad:
1g
25g
5g
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11319835

Marca: Thermo Scientific Alfa Aesar L19842.14

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Amino-2,6-dimethylpyridine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3430-33-9
Molekylformel C7H10N2
Molekylvikt (g/mol) 122.171
MDL-nummer MFCD01646409
InChI-nyckel WISXXOGOMDYNSN-UHFFFAOYSA-N
Synonym 3-amino-2,6-dimethylpyridine, 3-amino-2,6-lutidine, 2,6-dimethyl-3-pyridylamine, 3-pyridinamine, 2,6-dimethyl, 3-amino-2,6-dimethyl pyridine, 2,6-dimethyl-3-aminopyridine, 2,6-dimethyl-5-aminopyridine, 2,6-dimethylpyridin-3-ylamine, 2,6-dimethyl-3-pyridinylamine, zlchem 951
PubChem CID 76964
IUPAC-namn 2,6-dimethylpyridin-3-amine
LEDER CC1=NC(=C(C=C1)N)C

Especificaciones

Kemiskt namn eller material 3-Amino-2,6-dimethylpyridine
Smältpunkt 121°C to 123°C
Kokpunkt 230°C
Kvantitet 25 g
UN-nummer UN2811
Beilstein 2424
Löslighetsinformation Insoluble in water.
Formel vikt 122.17
Procent renhet 97%
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.