Learn More
Abnova™ Fluorescent Dye 649-C5 Alkyne
Cyanine 5 dyes are common red fluorophores, and can tolerate a pH range of 3 to 10; C5 Alkyne reacts with azide groups
Brand: Abnova™ U0211.1mg
Additional Details : Weight : 0.00010kg
Description
Ideal for labeling of biological molecules (e.g. peptides, proteins, oligoes) for fluorescence imaging and other fluorescence-based biochemical analysis
- Chemical Formula: C38H44F3N3O9S2
- Mol. Weight: 807.9g/mol
- Purity: ≥90%
- Molar Extinction Co-efficient: ≥1,00,000 M-1cm-1
- Excitation (Max): 649nm
- Emission (Max): 665nm
- Solubility: DMSO, water
Application: Conjugation, In vivo imaging
Recommended Storage: Store in the dark at -20°C.
Specifications
Cyanine 5 dyes are common red fluorophores, and can tolerate a pH range of 3-10. This C5 Alkyne reacts with azide groups. | |
1 mg | |
807.9g/mol | |
Store in the dark at -20°C | |
665 |
Conjugation; in vivo Imaging | |
Fluorescent Dye 649-C5 Alkyne | |
≥90% | |
Fluorescent Dye 649-C5 Alkyne | |
649 |
For research use only (RUO).