Learn More
Epredia™ Accessories for all Richard-Allan Scientific™ Cryostat Models, Cryostat oil
Support and expand the functionality of Epredia™ Cryostat Models with Accessories for all Epredia Cryostat Models.
Brand: Epredia™ 350040
438.60 SEK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Accessories Include:
Specifications
Cryostat Oil, 100mL | |
Cryostat Oil | |
Epredia Cryostats |