Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Supor™ Membrane
AcroPak capsules designed for research applications only.
Brand: Cytiva (formerly Pall Lab) 12093
Additional Details : Weight : 0.22900kg
Description
- Efficient processing of liquid volumes up to 20 L
- Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifications
Hydrophilic PES | |
Sterile | |
1/4 in. MNPT Inlet, Stepped Hose Barb Outlet |
0.8/0.2 μm | |
200 cm2 | |
3/Pk. |