missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ACVR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141185
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
By Western blot, this antibody detects a ~54 kDa band representing Activin Receptor Type IA. Additional non-specific bands at ~14 and 27 kDa may also be detected in human kidney samples. Suggested positive control: human kidney protein or tissue.
Activin A receptor, type I (ACVR1) belongs to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. ACVR1 signals a specific transcriptional response in concert with activin type II receptors. The ACVR1 gene encodes activin A type I receptor, which signals a particular transcriptional response in concert with activin type II receptors. Mutations in ACVR1 have been linked to Fibrodysplasia Ossificans Progressiva (FOP), a rare genetic disorder characterized by the formation of bone in muscles, tendons, and other connective tissues, as well as Epicanthus.
Specifikationer
| ACVR1 | |
| Polyclonal | |
| Unconjugated | |
| ACVR1 | |
| activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11477, 79558, 90 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| P37172, P80201, Q04771 | |
| ACVR1 | |
| A synthetic peptide corresponding to amino acids 475-525 of human ACVR1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering