missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Description
- Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
Specifications
Color | White |
Physical Form | Powder |
Quantity | 1 g |
Synonym | adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt |
IUPAC Name | disodium trihydrate hydrogen [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate |
Formula Weight | 551.2 (Anhydrous) |
Chemical Name or Material | Adenosine-5'-Triphosphate Disodium Salt Hydrate, Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'