Learn More
AIMS™ ATS-3 General Rodent Tattoo System Needles
Developed to minimize animal pain and tissue damage during tattooing
Specifications
For Use With (Application) Tattooing
Type Tattoo Needle
For Use With (Equipment) ATS-3 general rodent tattoo system
Material 401 Stainless Steel, Silver Solder
Sterility Sterile
View More Specs
Products 1
Product Code Brand Tip Style Price Quantity & Availability  
Product Code Brand Tip Style Price Quantity & Availability  
13498789
Encompass
View Documents
AIMS™
3035
Ultra Fine Point N/A
View alternative products
Save Up to 
»
N/A
Description
Description
Accessory for ATS-3 General Rodent Tattoo System
  • Made of 401 stainless steel and silver solder
  • Pre-sterilized and ready for tattooing
  • Proper needles allows for humane and rapid animal tattooing
  • Tails are not damaged
  • 5 needles per pack
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
Tattooing
ATS-3 general rodent tattoo system
Sterile
Tattoo Needle
401 Stainless Steel, Silver Solder