Learn More
3M™ Aircare™ Filtration Unit ACU-03
3M™ Aircare™ Filtration Unit ACU-03 provides high quality air from an external source of compressed air by removing water and oil droplets, solid particles, and odors.
Brand: 3M™ 7100035217
6545.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.10000kg
Description
- Supplies the highest quality air to the Supplied Air Respirator System and compressed air tools by removing water and oil aerosols, particles and odors from the facility supplied compressed air
- Wall mounted unit
- Can be used for up to three users or two users plus air tool
Specifications
CE Approved | |
ACU-03 | |
Wall-mounted Aircare Filtration Unit |
Gray | |
0.333 x 0.233 x 0.18 m |