Learn More
3M™ Aircare™ Filtration Unit ACU-04
3M™ Aircare™ Filtration Unit ACU-04 provides high quality air from an external source of compressed air by removing water and oil droplets, solid particles, and odors.
Brand: 3M™ 7100035348
6665.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.50000kg
Description
- Supplies the highest quality air to the Supplied Air Respirator System and compressed air tools by removing water and oil aerosols, particles and odors from the facility supplied compressed air
- Free standing unit
- Can be used for up to three users or two users plus air tool
Specifications
CE Approved | |
ACU-04 | |
Floor-mounted Aircare Filtration Units |
Light Gray | |
0.433 x 0.315 x 0.255 m |