Learn More
R&L Enterprises™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. R&L Enterprises™ Bossheads come with offset jaws for greater strength.
Brand: R&L Enterprises™ Z0204
Additional Details : Weight : 0.12060kg
Description
- Aluminum alloy with clamping screws
- For use with rods up to 16mm in diameter
Specifications
Bosshead Clamp | |
Alloy | |
with Blue Plastic headed nickel plated steel screws | |
For use with rods up to 16mm diameter |