missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ALOX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552728
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: YAWVPNAPCT MRMPPPTTKE DVTMATVMGS LPDVRQACLQ MAISWHLSRR QPDMVPLGHH KEKYFSGPKP KAVLNQFRTD LEKLEKEITA RNEQLDWPYE Y Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
The ALOX12 gene encodes the enzyme arachidonate 12-lipoxygenase, a member of the lipoxygenase family that metabolizes arachidonic acid into bioactive lipid mediators. ALOX12 catalyzes the formation of 12-hydroperoxyeicosatetraenoic acid (12-HPETE), which is further reduced to 12-hydroxyeicosatetraenoic acid (12-HETE). These products are involved in various biological processes, including inflammation, cellular signaling, and blood platelet function. ALOX12 plays a significant role in pathophysiological conditions such as cancer, cardiovascular diseases, and inflammatory disorders. Dysregulation or altered expression of ALOX12 has been implicated in tumorigenesis and metastasis, where its metabolites influence cell proliferation, migration, and survival. Additionally, ALOX12 impacts platelet aggregation and inflammation, making it a potential target for therapeutic strategies aimed at treating diseases linked to lipid mediator imbalances. Research into ALOX12 continues to explore its functions in health and disease, emphasizing its importance in lipid metabolism and signaling pathways.
Specifikationer
| ALOX12 | |
| Polyclonal | |
| Unconjugated | |
| ALOX12 | |
| 12(S)-lipoxygenase; 12LO; 12-LO; 12LOX; 12-LOX; 12S LOX; 12S-lipoxygenase; 12S-LOX; 9930022G08Rik; ALOX12; Alox12p; arachidonate 12-lipoxygenase; arachidonate 12-lipoxygenase, 12S type; arachidonate 12-lipoxygenase, 12S-type; Arachidonate 15-lipoxygenase,15S-type; Linoleate 13S-lipoxygenase; Lipoxin synthase 12-LO; LOG12; P-12LO; platelet 12-LOX; platelet-type 12-lipoxygenase; platelet-type lipoxygenase 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 239 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18054 | |
| ALOX12 | |
| Recombinant protein corresponding to Human ALOX12. Recombinant protein control fragment (Product #RP-90628). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering