missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ alpha Catenin Monoclonal Antibody (alpha-CAT-7A4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ 139700
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
alpha Catenin Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC, IP, Western Blot
Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.
Specifikationer
| alpha Catenin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P26231, P26232, P30997, P35221, Q61301 | |
| CTNNA1, CTNNA2 | |
| Synthetic peptide derived from the C-terminus of mouse alpha-catenin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Chicken, Frog | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| alpha-CAT-7A4 | |
| Unconjugated | |
| CTNNA1 | |
| 102 kDa cadherin-associated protein; 2010010M04Rik; AA517462; AI481747; AI988031; alpha E catenin; alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; alpha-catenin-related protein; alpha-E-catenin; alphaE-catenin; alpha-N-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; CAPR; CAP-R; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin associated protein), alpha 2; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha1; catenin alpha-1; Catenin alpha2; Catenin alpha-2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; Catna; Catna1; Catna2; cdf; cerebellar deficient folia; chp; CT114; CTNNA1; Ctnna2; CTNR; FLJ36832; FLJ52416; HGNC:2509; MDPT2; Neural alpha-catenin; NY-REN-13 antigen; renal carcinoma antigen NY-REN-13 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 12385, 12386, 1495, 1496, 297357, 307505, 396035, 416184 | |
| -20°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering