missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ Ammonia and Ammonium Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 251998
This item is not returnable.
View return policy
Description
Specifications
Tannin | |
For Photoflex™ photometers | |
0 to 50 mg/L NH4-N, 0.5 to 64.4 mg/L NH4+ | |
Photometric |
For waste water, drinking water, sea water | |
200 | |
200 Tests | |
CombiCheck Sulfate (SO4) reagent test kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Vario Ammonia Cyanurate F5 ml
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide, sodium dichloroisocyanurate, dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction