Learn More
STEMCELL Technologies Anacardic Acid
Epigenetic modifier; Inhibits histone acetyltransferase (HAT)
Brand:  STEMCELL Technologies 73194
Encompass_Preferred
 View more versions of this product
Product Code. 17180182
  • POA / 5mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Anacardic Acid is a 6-alkyl salicyclic acid that inhibits the histone acetyltransferase (HAT) activity of the transcription co-activators p300 and p300/CREB-binding protein-associated factor (pCAF; IC50 values of 8.5 and 5 μM, respectively; Balasubramanyam et al.) Anacardic Acid inhibits HAT-dependent transcription and protein SUMOylation (Cui et al.; Fukuda et al.) In addition, Anacardic Acid is an activator of Aurora kinase A-mediated phosphorylation of Histone H3 (Kishore et al.) At higher concentrations (25 μM), Anacardic Acid suppresses NF-κB activation and inhibits IκB-α phosphorylation (Sung et al.)
Specifications
Specifications
16611-84-0
5 mg
C22H36O3
≥98%