Läs mer
anti-Erythroid Cell Marker/TER-119 Antigen PE, Clone: TER-119, Invitrogen™
Erythroid Cell Marker/TER-119 Antigen Monoclonal antibody A18417 from Invitrogen specifically detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. It is validated for Flow Cytometry.
Brand: Invitrogen A18417
Beskrivning
Erythroid Cell Marker/TER-119 Antigen Monoclonal antibody A18417 from Invitrogen specifically detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. It is validated for Flow Cytometry.
TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG2b, κ
Clone: TER-119
Immunogen: Mouse Erythroid Cells.
Formulation: 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
This product detects Erythroid Cell Marker/TER-119 Antigen in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Mouse Erythroid Cells.
Specifikationer
| Erythroid Cell Marker/TER-119 Antigen | |
| Monoclonal | |
| PE | |
| Rat | |
| 50 μg | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| TER-119 | |
| 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide | |
| Mouse Erythroid Cells. | |
| Primary | |
| Store at 4° C | |
| IgG2b κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.