missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ AGFG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541784
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide sequence: SQSPVVGRSQ GQQQEKKQFD LLSDLGSDIF AAPAPQSTAT ANFANFAHFN Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
HRB is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. HRB binds the activation domain of the human immunodeficiency virus Rev protein when Rev is assembled onto its RNA target, and is required for the nuclear export of Rev-directed RNAs. The protein encoded by this gene is related to nucleoporins, a class of proteins that mediate nucleocytoplasmic transport. This encoded protein binds the Rev activation domain when Rev is assembled onto its RNA target and can significantly enhance Rev activity when overexpressed. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifikationer
| AGFG1 | |
| Polyclonal | |
| Unconjugated | |
| AGFG1 | |
| 0710001B06Rik; AGFG1; Agfg1-ps1; AI838505; arf-GAP domain and FG repeat-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeats-containing protein 1; arf-GAP domain and FG repeat-containing protein 1; ArfGAP with FG repeats 1; AU045498; C130049H11Rik; C85612; D730048C23Rik; HIV-1 Rev binding protein; HIV-1 Rev-binding protein; HIV-1 Rev-binding protein homolog; Hrb; hRIP, Rev interacting protein; MGC116938; MGC116940; NEUG(55-78); Neurogranin; neurogranin (protein kinase C substrate, RC3); Ng; NG/RC3; Nrgn; Nucleoporin-like protein RIP; postsynaptic spine 1; protein kinase C substrate; Pss1; R75334; RAB; Rab, Rev/Rex activation domain-binding protein; RC3; rev/Rex activation domain-binding protein; Rev-interacting protein; Rip; Unknown (protein for MGC:128795) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3267 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P52594 | |
| AGFG1 | |
| Synthetic peptide directed towards the middle region of human HRB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering