Learn More
anti-IGF1R, PIK3R3 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0268.1Set
Additional Details : Weight : 0.00010kg
Includes: IGF1R rabbit purified polyclonal antibody (20μg); PIK3R3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the IGF1R protein, and the other against the PIK3R3 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
IGF1R | |
Monoclonal/Polyclonal | |
IGF1R | |
IGF1R | |
1 Set | |
Primary | |
Antibody pair set content: 1. IGF1R rabbit purified polyclonal antibody (20 ug) 2. PIK3R3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
CD221/IGFIR/JTK13/MGC142170/MGC142172/MGC18216 | |
Mouse, Rabbit | |
RUO | |
3480/8503 | |
Human | |
IGF1R & PIK3R3 Protein Protein Interaction Antibody Pair |