missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
IL1RN, Human, Matched Antibody Pair (AP22), Abnova™
To detect and quantify protein level of human
Brand: Abnova H00003557-AP22.1Set
Innehåller: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-IL1RN (100μg) , Detection antibody: mouse purified polyclonal anti-IL1RN (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifikationer
| IL1RN | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL1RN. | |
| ICIL-1RA/IL-1ra3/IL1F3/IL1RA/IRAP/MGC10430 | |
| Mouse, Rabbit | |
| RUO | |
| 3557 | |
| Human | |
| Liquid |
| ELISA | |
| Unconjugated | |
| IL1RN | |
| IL1RN | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IL1RN (100 μg) 2. Detection antibody: mouse purified polyclonal anti-IL1RN (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering