Läs mer
Invitrogen™ Interferon alpha-6 Monoclonal Antibody (3C9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA519186
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Peptide Sequence: WDERLLDKLY TELYQQLNDL EACVMQEVWV GGTPLMNEDS ILAVRKYFQR ITLYLTEKKY SPCAWEVVRA EIMRSFSSSR NLQERLRRKE.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifikationer
| Interferon alpha-6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P05013 | |
| IFNA6 | |
| IFNA6 (NP_066282, 100 a.a. ~ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 3C9 | |
| Unconjugated | |
| IFNA6 | |
| IFNA6; IFN-alpha-6; IFN-alphaK; Interferon; interferon alpha 6; interferon alpha-54; Interferon alpha-6; interferon alpha-K; interferon, alpha 6; leIF K; RP11-354P17.7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3443 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |