Learn More
anti-PITX2, CTNNB1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0117.1Set
Additional Details : Weight : 0.00010kg
Includes: PITX2 rabbit purified polyclonal antibody (20μg); CTNNB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PITX2 protein, and the other against the CTNNB1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
PITX2 | |
Monoclonal/Polyclonal | |
PITX2 | |
PITX2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. PITX2 rabbit purified polyclonal antibody (20 ug) 2. CTNNB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
ARP1/Brx1/IDG2/IGDS/IGDS2/IHG2/IRID2/MGC111022/MGC20144/Otlx2/PTX2/RGS/RIEG/RIEG1/RS | |
Mouse, Rabbit | |
RUO | |
5308/1499 | |
Human | |
PITX2 & CTNNB1 Protein Protein Interaction Antibody Pair |