missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
anti-RAD51, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00005888-PW1.1Set
Innehåller: Antibody pair for IP: mouse monoclonal anti-RAD51 (300μg), Antibody pair for WB: rabbit polyclonal anti-RAD51 (50μL)
Specifikationer
| RAD51 | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| BRCC5/HRAD51/HsRad51/HsT16930/RAD51A/RECA | |
| Mouse, Rabbit | |
| RUO | |
| 5888 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RAD51 | |
| RAD51 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-RAD51 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-RAD51 (50 ul) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering