Learn More
BINDER™ APT-COM™ 4 Series
Multi Management Software provides management, logging, programming, and documentation options. With features ranging from GLP-compliant logging to convenient program creation and management of up to 100 units, the APT-COM™ 4 can be used in all industries and makes complex work processes much easier.
Brand:  BINDER™ 9053-0039
 View more versions of this product
Product Code. 16307175
  • 3225.00 SEK / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Description
APT-COM™ 4 BASIC Edition:
  • Administration of up to five connected units
  • Log management (creating, deleting, archiving)
  • Documentation of measured values
  • Central overview of all units in both diagram and table form
  • Graphical presentation of measured values
  • Graphical/numerical program editor
  • Manual export of measured values (CSV/PDF file)
  • Multilingual user interface (German, English, French, Spanish, Italian)
  • Optional program execution via APT-COM™
  • Timer function
  • Import of data from APT-COM™ 3
APT-COM™ 4 PROFESSIONAL Edition:
All the same functions as the BASIC Edition and:
  • Administration of up to 100 connected units at the same time
  • Central overview of all units in a room plan
  • Email-based alarm system
  • Web server for displaying results
  • User administration for multiple users and authorizations
  • Automatic, time-managed export of measured values
  • Reports for further processing/archiving
  • Automatic/manual backup and recovery
APT-COM™ 4 GLP Edition
All the same functions as the PROFESSIONAL Edition and:
  • Can be validated according to 21 CFR Part 11 including validation documents
  • Documentation of measured values in a tamper-proof file in accordance with GLP/GMP guidelines on audit trails
  • Electronic signatures
For Use With: BINDER Chambers Series BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL
Specifications
Specifications
Data Collection Software
4 BASIC-Edition Software
BINDER Chambers Series (BD, BF, CB-S, CB, CBF, ED, FD, FED, FP, M, KB, KB ECO, KT, KBF, KBF-S, KBF-S ECO, KBF P, KBF LQC, KBW, KBWF, UFV, VD, VDL)
APT-COM™ 4 BASIC-Edition - for simple logging and documentation requirements with up to 5 networked units.
Logging, Controlling, Monitoring
APT-COM