Learn More
Cytiva BAS Storage Phosphor Screens
Use phosphor screens to capture latent images produced by ionizing radiation. Cytiva™ BAS Storage Phosphor Screens emit light upon laser-induced stimulation in proportion to the amount of radioactivity in the sample.
8064.00 SEK - 44511.00 SEK
Specifications
Quantity 1Each
View More Specs
Products 9
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11360312
View Documents
Cytiva
28-9564-75
BAS Storage Phosphor Screens
14060.00 SEK
Each
In Stock
to see stock.
Add to basket
11350312
View Documents
Cytiva
28-9564-74
BAS Storage Phosphor Screens
22403.00 SEK
Each
In Stock
to see stock.
Add to basket
11370312
View Documents
Cytiva
28-9564-76
BAS Storage Phosphor Screens
44511.00 SEK
Each
In Stock
to see stock.
Add to basket
11310322
View Documents
Cytiva
28-9564-80
BAS Storage Phosphor Screen
8064.00 SEK
Each
In Stock
to see stock.
Add to basket
11300322
View Documents
Cytiva
28-9564-79
BAS Storage Phosphor Screens This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11380312
View Documents
Cytiva
28-9564-77
BAS Storage Phosphor Screens
28235.00 SEK
Each
In Stock
to see stock.
Add to basket
11320322
View Documents
Cytiva
28-9564-81
BAS Storage Phosphor Screens
15105.00 SEK
Each
Estimated Shipment: 13-12-2024
to see stock.
Add to basket
10232628
View Documents
Cytiva
28-9564-82
BAS Storage Phosphor Screens
10020.00 SEK
Each
In Stock
to see stock.
Add to basket
11390312
View Documents
Cytiva
28-9564-78
BAS Storage Phosphor Screens
18226.00 SEK
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Requires approximately one-tenth the exposure time of traditional autoradiography film.
  • Ten to one hundred times more sensitive than film, depending on isotope and sample type.
  • Wide linear dynamic range (five orders of magnitude) enables quantitation and visualization of both weak and strong signals in a single exposure.
  • Screens do not require chemicals, darkroom, or other special treatment for use.
  • Each step can be performed at the lab bench under normal lighting conditions and at room temperature.
  • Results are digitized using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software.
Specifications
Specifications
1Each