Learn More
Benzyl chloroformate, 3M (50 wt%) solution in toluene, Thermo Scientific Chemicals
433.40 SEK - 1619.55 SEK
Specifications
Chemical Name or Material | Benzyl chloroformate |
---|---|
Name Note | 50 wt. % Solution in Toluene |
CAS | 501-53-1, 108-88-3, 100-44-7 |
CAS Min % | 45.0, 47.0 |
CAS Max % | 53.0, 55.0 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773904
|
Thermo Scientific Acros
124491000 |
100 mL | Glass bottle |
433.40 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10265410
|
Thermo Scientific Acros
124495000 |
500 mL | Glass bottle |
1619.55 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
Benzyl chloroformate | |
501-53-1, 108-88-3, 100-44-7 | |
53.0, 55.0 | |
7°C | |
C8H7ClO2 | |
MFCD00000640 | |
01,109; 02,59; 15,22 | |
1.01 | |
Solubility in water: decomposes | |
C1=CC=C(C=C1)COC(=O)Cl | |
170.6 | |
170.6 | |
Solution |
50 wt. % Solution in Toluene | |
45.0, 47.0 | |
1.0100g/mL | |
Glass bottle | |
C6H5CH2OOCCl | |
06, 437 | |
15, 1801 | |
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester | |
HSDAJNMJOMSNEV-UHFFFAOYSA-N | |
benzyl carbonochloridate | |
10387 | |
45 to 53 wt% |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness. May cause cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Suspected of damaging the unborn child. Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Avoid release to the environment.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-925-0
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Carcinogenicity Category 1B
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H304-May be fatal if swallowed and enters airways.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Toluene, Benzyl chloroformate, Benzyl chloride
RUO – Research Use Only