Learn More
(Benzylthio)acetone, 98%, Thermo Scientific Chemicals
Product Code. 11336666
Click to view available options
Quantity:
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10230-69-0
Molecular Formula C10H12OS
Molecular Weight (g/mol) 180.265
MDL Number MFCD00026241
InChI Key OIEDQMIEPJIRFT-UHFFFAOYSA-N
Synonym 1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
PubChem CID 82472
IUPAC Name 1-benzylsulfanylpropan-2-one
SMILES CC(=O)CSCC1=CC=CC=C1
Specifications
Density 1.09
Boiling Point 155°C to 156°C (17 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.558
Quantity 5 g
Beilstein 1866268
Formula Weight 180.27
Percent Purity 98%
Chemical Name or Material (Benzylthio)acetone