missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Black Urea Screw Caps with Wood Pulp Faced PVDC Liner
Click to view available options
Closure Size:
100/R3
22/R3
24/R3
28/R3
31/R6
33/R3
38/R3
48/R3
51/R3
58/R3
63/R3
83/R3
Description
- Black urea plastic
- Includes fitted wood pulp faced PVDC liner
- Supplied in packs of 50
Specifications
Specifications
Color | Black |
Material | Urea |
Quantity | 50/PK |
Liner Material | Wood pulp faced PVDC |
For Use With (Equipment) | Wide mouth bottles and jars |
Closure Size | 100/R3 |
Closure Type | Screw Cap |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'