missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Blood Agar Base
Blood Agar Base is a nutrient rich medium, suitable for the isolation of pathogenic microorganisms from clinical specimens.
Brand: Scharlab 01-352-500
This item is not returnable.
View return policy
Description
- Nutrient rich medium suitable for the isolation of pathogenic microorganisms from clinical specimens.
- This medium needs to add Ampicillin Selective Supplement (Art.No.: 06-126LYO1).
- This medium needs addition of Blood.
Specifications
Red | |
Blood Agar Base | |
Meat extract 10.00 g/L, Tryptone 10.00 g/L, Sodium chloride 5.00 g/L, Agar 15.00 g/L | |
37°C ± 1.0°C | |
7.3 ±0.2 | |
Insoluble |
Solid | |
Microbiology Media | |
Isolation of Pathogenic Microorganisms | |
24 to 48 hr. | |
500 g |